Corning® 1536 well cyclic olefin copolymer (COC) microplate
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4560 4561 4563 4564 4565 4571 4572
CORN4560EA 2474.3 EUR
CORN4560 CORN4561 CORN4563 CORN4564 CORN4565 CORN4571 CORN4572
Corning® 1536 well cyclic olefin copolymer (COC) microplate
Microplaten Celcultuurplaten
Cyclic olefin copolymer (COC). Superior flatness of the plates minimises variability within a plate and between plates. The barcoded plates come with with raised well bottoms for higher sensitivity.
  • Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
  • No logo or lettering for improved heat sealing and ease of liquid handling
  • Working volume is up to 10 µl
  • Flat bottom wells are compatible with automated dispensers