Evsco Valves Manual and Actuated Accessories
Leverancier: Evsco
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MFLX98150-50EA 27.1 EUR
MFLX98150-50 MFLX98150-51
Evsco Valves Manual and Actuated Accessories
Ventielen Elliptical Valves
Safely mount your plastic elliptical valves.
  • Optimize performance of your Evsco Valves
Constucted of durable polypropylene, these mounting brackets are ideal for making your valve more versitile. These mounting brackets allow for wall, panel, or undermount.