Masterflex® Reducer Fittings, Hose Barb, Tee, Avantor®
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
31702-45 31702-47 31702-57 31702-61 31702-62
MFLX30401-54EA 182 EUR
MFLX30401-54 MFLX30401-55 MFLX30401-56 MFLX30401-58 MFLX30401-60 MFLX30401-63 MFLX31208-41 MFLX31702-45 MFLX31702-47 MFLX31702-57 MFLX31702-61 MFLX31702-62 MFLX30401-59 MFLX31208-42 MFLX31208-44 MFLX31208-45 MFLX31208-46 MFLX31208-47 MFLX31208-48 MFLX40615-48 MFLX40614-36 MFLX40615-50 MFLX40615-51
Masterflex® Reducer Fittings, Hose Barb, Tee, Avantor®
Fittingen en connectoren
Quick and easy assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).