Masterflex® Union Fittings, Hose Barb, Cross Connector, Avantor®
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MFLX40726-70EA 231 EUR
MFLX40726-70 MFLX40726-71 MFLX40726-73 MFLX40726-74 MFLX31208-54 MFLX31407-11 MFLX31408-11 MFLX31208-51 MFLX31208-52 MFLX31208-53 MFLX31208-55 MFLX40615-83 MFLX40615-84
Masterflex® Union Fittings, Hose Barb, Cross Connector, Avantor®
Fittingen en connectoren
Quick and easy assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed fittingsMore Product Information