PFA Straight Threaded Couplings, NPT (F)
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
06546-00 06546-01 06546-02 06546-03 06546-04
MFLX06546-00EA 48.3 EUR
MFLX06546-00 MFLX06546-01 MFLX06546-02 MFLX06546-03 MFLX06546-04
PFA Straight Threaded Couplings, NPT (F)
Fittingen en connectoren
Create a secure pipe threaded connection.
  • Constructed of chemically inert PFA
These straight threaded coupling fittings are made of chemically inert PFA. Fittings can be sterilized via autoclave or ethylene oxide (ETO).