Reducer Fittings, Threaded Bushing, Straight
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
06546-05 06546-06 06546-07 06546-08 06546-09 06546-10 34030-43 34030-44 34030-45 34030-46 34030-50 34030-54
MFLX06546-05EA 150 EUR
MFLX06546-05 MFLX06546-06 MFLX06546-07 MFLX06546-08 MFLX06546-09 MFLX06546-10 MFLX34030-43 MFLX34030-44 MFLX34030-45 MFLX34030-46 MFLX34030-50 MFLX34030-54
Reducer Fittings, Threaded Bushing, Straight
Fittingen en connectoren
Maintain a permanent connection.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Threaded fittings offer more permanent installation. Choose the size and material that meets your application needs.