Liquid or Gas Turbine Flowmeters / Transmitters
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
32709-02 32709-08 32709-10 32709-12 32709-14 32709-50 32709-52 32709-54 32709-56 32709-58 32709-60 32709-70 32709-72 32709-74 32709-78
MFLX32709-02EA 739 EUR
MFLX32709-02 MFLX32709-08 MFLX32709-10 MFLX32709-12 MFLX32709-14 MFLX32709-50 MFLX32709-52 MFLX32709-54 MFLX32709-56 MFLX32709-58 MFLX32709-60 MFLX32709-70 MFLX32709-72 MFLX32709-74 MFLX32709-78
Liquid or Gas Turbine Flowmeters / Transmitters
Debietmeters
Output signal lets you connect to a remote display, datalogger, or recorder for continuous monitoring.
  • The 3-1/2 digit LCD provides direct flow rate readings
These new low-flow liquid and gas flowmeters are ideal for industrial, commercial, laboratory, or OEM applications. They are compact and offer excellent liquid or gas measurements. All models provide direct flow rate readings in ml/min or L/min and a 0 to 5 VDC linear output simultaneously.
Choose from RYTON® or brass flowmeters. RYTON meters are an economical alternative to brass models. OrderMore Product Information