Masterflex® Standard Pump Head Tubing Loading Keys, Avantor®
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MFLX07013-90EA 10.5 EUR
MFLX07013-90 MFLX07019-90
Masterflex® Standard Pump Head Tubing Loading Keys, Avantor®
Uittrek- en invoeggereedschap
Ensure proper tubing alignment and installation.
  • Constructed of durable stainless steel
L/S® and I/P® Standard pump heads include tubing loading keys. A key is required to ensure proper tubing alignment and tension. Order a replacement key based on your pump head series, L/S® or I/P®.