Gast Direct-Drive NEMA Type 56 C-Face Motors
Leverancier: Gast
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MFLX07055-52EA 574 EUR
MFLX07055-52 MFLX07055-41 MFLX07055-76
Gast Direct-Drive NEMA Type 56 C-Face Motors
Pompen Motors
Four-vane rotary design is almost maintenance free.
  • Nonsparking, nonelectrical operation—safe and reliable
These direct drive air motors feature a 350 to 3500 rpm speed range and a maximum pressure rating of 100 psi.
Leveringsinformatie: Includes motors come with an exhaust muffler. Install a compressed-air filter in-line to remove oil, water, and solid contaminants—order connecting nipples and fliters separately.