Viton® Transfer Pump Tubing
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MFLX06434-01EA 69.7 EUR
MFLX06434-01 MFLX06434-02 MFLX06434-03 MFLX06434-04 MFLX06434-05 MFLX06434-06 MFLX06434-07 MFLX06434-08 MFLX06434-09
Viton® Transfer Pump Tubing
Slangen Transfer Tubing
keep light sensitive samples safe.
  • Global market leader in range of formulations for most extensive chemical compatibility
Applications: Acid and solvent transfer; high-temperature work.
Characteristics: Opaque black.
Certification: Tubing is manufactured according to GMP.
Temperature range: –25 to 399 °F (–32 to 204 °C)
Sterilize: Not recommended.