Masterflex® Transfer Tubing, Perfluoroalkoxy (PFA), Avantor®
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
06371-07 06371-08 06371-22 06371-23
MFLX06371-00EA 62.5 EUR
MFLX06371-00 MFLX06371-01 MFLX06371-02 MFLX06371-03 MFLX06371-04 MFLX06371-05 MFLX06371-06 MFLX06371-21 MFLX06371-24 MFLX06371-25 MFLX06371-26 MFLX06371-28 MFLX06371-29 MFLX06375-01 MFLX06375-02 MFLX06375-04 MFLX06375-05 MFLX06375-07 MFLX06375-08 MFLX06371-07 MFLX06371-08 MFLX06371-22 MFLX06371-23
Masterflex® Transfer Tubing, Perfluoroalkoxy (PFA), Avantor®
Slangen Transfer Tubing
Stands up to highly corrosive materials.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
Applications: Demanding lab, chemical, electrical, mechanical, pharmaceutical, and automotive applications. For the transfer of high-temperature or highly corrosive materials. Also good for high-purity work.
Characteristics: Chemically inert, nontoxic, and virtually nonporous. Excellent crack- and stress-resistance. Low coefficient of friction. Translucent.
Resin certification: Resin is FDA-compliant (21 CFR 177.1550); meets USP Class VI requirements. Tubing is manufactured according to GMP.
Temperature range: -320 to 500 °F (-195 to 260 °C)
Sterilize: Autoclave, ethylene oxide, or use dry heat.