Double-Walled Heat-Shrink Tubing, PTFE
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
95810-13 95810-15 95810-16
MFLX95810-13EA 495 EUR
MFLX95810-13 MFLX95810-15 MFLX95810-16
Double-Walled Heat-Shrink Tubing, PTFE
Slangen Heat-Shrink Tubing
Ideal for chemically inert applications.
  • Global market leader in range of formulations for most extensive chemical compatibility
Double-walled PTFE tubing combines an exterior of heat-shrink PTFE with an inner layer of FEP to provide a tight, moisture-proof bond. Comes in 4-ft lengths; 3 lengths per pack, for a total of 12 ft/pk. Opaque.