Miniature Plastic Hose Barb Check Valves
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MFLX98553-00EA 7.1 EUR
MFLX98553-00 MFLX98553-11 MFLX98553-22 MFLX98553-01 MFLX98553-12 MFLX98553-34 MFLX98553-20 MFLX98553-21 MFLX98553-10 MFLX98553-13 MFLX98553-02
Miniature Plastic Hose Barb Check Valves
Ventielen Check Valves
Use with liquids and gases for pressure or vacuum applications.
  • Breaking pressure is 0.18 psi, closing pressure is 0.014 psi
  • Install in any position
Note: Valves require 1 psi back pressure for a tight seal.