GF Signet Needle Valves with Thread Adapters, PVDF
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
01369-21 01369-22
MFLX01369-21EA 560 EUR
MFLX01369-21 MFLX01369-22
GF Signet Needle Valves with Thread Adapters, PVDF
Ventielen Needle Valves
Create a secure pipe threaded connection.
  • Constructed of durable PVDF
PVDF needle valves are pressure rated up to 200 psi (13.8 bar) with a max temperature rating of 302 deg F (150 deg C). Seats are made of PTFE and O-rings made of Viton®. Valves feature standard pipe thread connections and are autoclavable.