Polypropylene and PVC Pressure Regulators
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MFLX01350-82EA 948 EUR
MFLX01350-82 MFLX01350-86
Polypropylene and PVC Pressure Regulators
Ventielen Overdrukventielen
Easily control your outlet pressure.
  • Choose between PVC or polypropylene (PP) construction with Viton® diaphragm
Valves feature a diaphragm made of Viton®. Use to control your outlet pressure between 5 and 50 psig; up to 150 psig inlet. Ideal for your liquid applications.