CPC® Plastic Quick-Disconnect Fittings, Hose Barb Inserts
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MFLX31311-61EA 29.2 EUR
MFLX31311-61 MFLX31311-62 MFLX31311-63 MFLX31311-71 MFLX31311-72 MFLX31311-73 MFLX06360-93 MFLX31311-73BLK MFLX06360-92BLK MFLX06360-93BLK MFLX31311-61BLK MFLX31311-71BLK MFLX31311-63BLK MFLX31311-62BLK MFLX31311-72BLK MFLX61027-97BLK
CPC® Plastic Quick-Disconnect Fittings, Hose Barb Inserts
Fittingen en connectoren
Fittings connect in a snap.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Acetal and ABS fittings are ideal for use with corrosive fluids
  • Polycarbonate and polysulfone materials meet USP Class VI requirements
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. To disconnect fitting halves, simply press the button on the coupling body.
A complete coupler requires a fitting body and an insert (order separately).