CPC® Plastic Quick-Disconnect Fittings, Sealing Caps
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
31311-67 31311-67BLK
MFLX31311-64EA 27.2 EUR
MFLX31311-64 MFLX31311-74 MFLX31311-77 MFLX31311-65 MFLX31311-67 MFLX31311-67BLK MFLX31311-64BLK MFLX31311-77BLK MFLX31311-65BLK MFLX31311-74BLK
CPC® Plastic Quick-Disconnect Fittings, Sealing Caps
Fittingen en connectoren
Fittings connect in a snap.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Polycarbonate and polysulfone materials meet USP Class VI requirements
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. To disconnect fitting halves, simply press the button on the coupling body.
A complete coupler requires a fitting body and an insert (order separately).