CPC® Metal Quick-Disconnect Fittings, Threaded Bodies
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
31403-00 31403-01 31403-05 31404-00 31404-20 31404-21 31403-01BLK 31404-00BLK 31403-00BLK 31403-05BLK 31404-21BLK 31404-20BLK
MFLX31403-00EA 36.1 EUR
MFLX31403-00 MFLX31403-01 MFLX31403-05 MFLX31404-00 MFLX31404-20 MFLX31404-21 MFLX31403-01BLK MFLX31404-00BLK MFLX31403-00BLK MFLX31403-05BLK MFLX31404-21BLK MFLX31404-20BLK
CPC® Metal Quick-Disconnect Fittings, Threaded Bodies
Fittingen en connectoren
Convenience of a quick-disconnect in pressure and vacuum applications.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Constructed of durable chrome-plated brass
  • Easy one-hand connection and disconnection
These CPC® metal quick-disconnect fittings are ideal for pressure and vacuum applications. Fittings are made of chrome-plated brass with stainless steel springs and latch. Valved fittings have an acetal shutoff valve that seals the flow path when fitting halves are disconnected. Can withstand vacuum to 250 psi, temperatures from −40 to 180 °F, and can be used with gases or liquids.
A complete coupler requires a fitting body and an insert with the same flow size (order separately).