Masterflex® Polypropylene String-Wound Gradient-Density Cartridges, Avantor®
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Masterflex®
MFLX01513-04EA 19.2 EUR
MFLX01513-04 MFLX01513-00 MFLX01513-02
Masterflex® Polypropylene String-Wound Gradient-Density Cartridges, Avantor®
Filters Filterpatronen
String-wind pattern has a lower presure drop for higher flow.
  • Constructed of polypropylene (PP)
Ideal for removing a broad range of deformable and nondeformable particles such as sand, silt, sludge, rust, and scale. Polypropylene string is resistant to chemical and bacterial attack. Maximum temperature is 150 °F (66 °C).
Note: For optimal performance, change cartridge at 25 psid.