Masterflex® Transfer Tubing, Adapta Flex™, Avantor®
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MFLX95951-01EA 131 EUR
MFLX95951-01 MFLX95951-09 MFLX95951-03 MFLX95951-05 MFLX95951-07 MFLX95951-11
Masterflex® Transfer Tubing, Adapta Flex™, Avantor®
Slangen Transfer Tubing
Maintain flexibility at high pressure.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Outstanding resistance to heat and ozone
Masterflex® Adapta Flex™ transfer tubing is ideal for general purpose applications requiring maximum flexibility. The EPDM tube and tube cover along with the high tensile textile core tube reinforcement makes this tubing suitable for high pressure applications. The tube and tube cover meet RMA Class C limitedMore Product Information