Corning® Lymphocyte serum-free medium
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
88-581-CM 88-551-CM
MDTC88-581-CMEA 253.08 EUR
MDTC88-581-CM MDTC88-551-CM
Corning® Lymphocyte serum-free medium
Celcultuurmedia
The serum-free medium is used to support the growth of T-cells activated with immobilized anti-CD3 antibody. Interleukin-2 is a necessary component for a successful culture. This formulation has been shown to work with the following cell types: CIK/DC-CIK cell lines, T-cells, CAR-T, TCR-T and TIL.
  • DMF available
  • Buffering ability is strengthened to keep pH stable during culture
The serum-free medium is an updated version and is ideal for serum reduced or serum-free culture conditions. This medium is compatible with following: CIK cells, NK cells, CAR-NK lines, T-cells, CAR-T, TCR-T and TIL.
Both formulations are prepared using select high quality reagents and raw materials including GMP-grade material. Both media are manufactured in certified ISO 9001 and ISO 13485 facilities.
Leveringsinformatie: Shipped on ice packs.