MultiRack filter tips, Axygen®
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MRF-100-L-R-S MRF-100-R-S MRF-1000XT-LRS MRF-1000XT-R-S MRF-10XT-L-R-S MRF-10XT-R-S MRF-20-L-R-S MRF-20-R-S MRF-200NX-L-RS MRF-200NX-R-S MRF-200XT-L-RS MRF-200XT-R-S MRF-300XT-L-RS MRF-300XT-R-S
AXYGMRF-100-L-R-SEA 659 EUR
AXYGMRF-100-L-R-S AXYGMRF-100-R-S AXYGMRF-1000XT-LRS AXYGMRF-1000XT-R-S AXYGMRF-10XT-L-R-S AXYGMRF-10XT-R-S AXYGMRF-20-L-R-S AXYGMRF-20-R-S AXYGMRF-200NX-L-RS AXYGMRF-200NX-R-S AXYGMRF-200XT-L-RS AXYGMRF-200XT-R-S AXYGMRF-300XT-L-RS AXYGMRF-300XT-R-S
MultiRack filter tips, Axygen®
Pipettips
The Axygen® MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an interchangeable complete system offered in bulk, racked, filter and reload packaging styles to meet the variety of needs in the lab setting.
  • Detectable DNase/RNase/Human DNA/PCR inhibition free
  • Non pyrogenic
  • Manufactured in an ISO certified facility in the USA
Axygen® MultiRack filter tips employ an inert, hydrophobic barrier to prevent contamination from pipettors. The filter materials, containing no leachable additive with less than 20 µm pore sizes, effectively block aerosols without the risk of contaminating the precious sample with additives. Axygen® MultiRack filter tips are available in a wide range of tip choices.
Bestelinformatie: Axygen® MultiRack pipette tips are also available in bulk, hinged rack, reload, and tipstation configurations.