Sodium bicarbonate, Corning®
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
61-065-RR 61-065-RO 25-035-CI
MDTC61-065-RREA 235.92 EUR
MDTC61-065-RR MDTC61-065-RO 25-035-CI
Sodium bicarbonate, Corning®
Buffers Buffers voor weefselkweek
Sodium bicarbonate is used as part of a buffering system commonly used to maintain physiological pH 7,2 to 7,4 in a culture environment
Bicarbonate is a weak buffer with that makes maintaining a pH range of 7,2 to 7,6 somewhat difficult, but it is non-toxic, and is a nutrient source.