Multirack bulk tips, Axygen®
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MR-1000XT MR-10XT MR-200
AXYGMR-1000XTEA 390 EUR
AXYGMR-1000XT AXYGMR-10XT AXYGMR-200
Multirack bulk tips, Axygen®
Pipettips
The Axygen® MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an inter-changeable complete system offered in bulk, racked, filter and reload packaging styles to meet the variety of needs in the lab setting.
  • Detectable DNase/RNase/Human DNA/PCR inhibition free
  • Nonpyrogenic
  • Manufactured in an ISO certified facility in the USA
  • Fully autoclavable
Axygen® MultiRack bulk pipette tips are compatible with MultiRack racks and packaged in resealable polyethylene bags, non sterile.
Bestelinformatie: Axygen® MultiRackMore Product Information