Automation tips for Agilent®/Velocity11 VPrep® and Bravo workstations, Axygen®
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VT-30UL-R VTF-20-R-S VTF-165-R-S VT-20-R-LAB-S VT-50-R VT-384-70UL-RS VTF-384-50ULRS VT-20-R VT-250-R VT-250-R-S VT-250-R-LAB VT-250-R-LABS VT-384-10UL-R VT-384-10UL-RS VT-384-31UL-R VT-384-31UL-RS VT-384-5UL-R VT-384-70UL-R VTF-384-15ULRS VT-255-R VTF-205-R-S
736-0352EA 0 EUR
736-0352 AXYGVTF-20-R-S 613-1911 736-0544 AXYGVT-50-R AXYGVT-384-70UL-RS AXYGVTF-384-50ULRS AXYGVT-20-R AXYGVT-250-R AXYGVT-250-R-S AXYGVT-250-R-LAB AXYGVT-250-R-LABS AXYGVT-384-10UL-R AXYGVT-384-10UL-RS AXYGVT-384-31UL-R AXYGVT-384-31UL-RS AXYGVT-384-5UL-R AXYGVT-384-70UL-R AXYGVTF-384-15ULRS AXYGVT-255-R AXYGVTF-205-R-S
Automation tips for Agilent®/Velocity11 VPrep® and Bravo workstations, Axygen®
Pipettips
Axygen® automation tips are engineered to the exact specifications of the original equipment manufacturers’ tips, ensuring complete compatibility and a consistent fit.
  • Maxymum Recovery® version available in most tip styles
  • Filtered version available in most tip styles to prevent aerosol and liquid contamination
Axygen® offers a comprehensive line of tips for Agilent®/Velocity11 automation systems. These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.Available in nonsterile, sterile, non-filtered, filtered, and wide-bore formats. Also available in Maxymum Recovery® to further improve accuracy and precision while reducing sample loss. Clear polypropylene tips are available in 96-well and 384-well format designed for Agilent®/Velocity VPrep® and Bravo 96LT, 8LT, 384ST, 96ST and 16ST heads.
Certificaten: Tips are certified RNase-/DNase-free, and nonpyrogenic.