Permaflow irradiated cleanroom ballpoint pen, BioClean™
Leverancier: Nitritex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S-BPBP-1-BK S-BPBP-1-BL
NITRS-BPBP-1-BKEA 1750 EUR
NITRS-BPBP-1-BK NITRS-BPBP-1-BL
Permaflow irradiated cleanroom ballpoint pen, BioClean™
Pennen Cleanroompennen
BioClean™ Permaflow cleanroom pens have fade and water-resistant fast drying permanent ink and feature an alcohol-resistant label for easy cleaning within a critical environment.
  • Alcohol-resistant labelling
  • Processed to ensure EU GMP Grade A compatibility
  • Permanent ink
  • Fade- and water-resistant
  • Quick drying
  • Non-toxic
Lightweight, tinted polypropylene round barrel and 1,0 mm point size for a line width of 0,32 mm for quality writing precision. Overall length is 149,8 mm.
Certificaten: Processed in a NEBB certified ISO Class 4 cleanroom environment. In compliance with ISO 12757-1 and ISO 12757-2 standards.
Verpakking: One pen per inner PE bag; three pens per sealed outer PE bag; 10 packs of 3 pens per sealed outer PE bag (30 pens per outer); 10 outer PE bags of 30 pens per carton liner; one carton liner per carton (300 pens).