GoatThroat Spillproof Barrel Pumps, Polypropylene
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
75650-10 75650-12 75650-14 75650-16
MFLX75650-10EA 387 EUR
MFLX75650-10 MFLX75650-12 MFLX75650-14 MFLX75650-16
GoatThroat Spillproof Barrel Pumps, Polypropylene
Pompen Vatpompen
Pump wide selection of fluids from 5 gallon pails to 55 gallon barrels.
  • Choice of four seal materials to handle over 800 liquids and chemicals
  • Guaranteed for 1 year. Life expectancy 7–10 years
  • Internal safety relief valve prevents overpressurization
  • Drums are Resource Conservation and Recovery Act (RCRA) ready
The GoatThroat pumps are designed to handle a wide range of liquids—wetted parts are 100% nonreactive, food-grade polypropylene. Choose nitrile seals for petroleum-based products including oil, diesel fuel, and glycerin. The EPDM seals are idealMore Product Information