General-Purpose TEFC/ODP, NEMA Type C-Face Four-In-One Motors
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
71004-25 71005-15 71005-25
MFLX71004-25EA 659 EUR
MFLX71004-25 MFLX71005-15 MFLX71005-25
General-Purpose TEFC/ODP, NEMA Type C-Face Four-In-One Motors
Pompen Motors
Totally enclosed motors are ideal for dusty, dirty, or damp environments.
  • Unique four-in-one design allows for replacement of ODP, TEFC, C-Face, or rigid based motors
  • Use for vertical or horizontal applications
Single-phase motors include a removable, rigid base. Three-phase motors inlcude a removable base.