Masterflex® Tuf-Flex® Sanitary Tri-Clamp® Gaskets, Avantor®
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
30547-60_U 30547-58
MFLA30547-60_UEA 0 EUR
MFLA30547-60_U MFLX30547-58 MFLX00278-GL MFLX30547-50 MFLX30547-52 MFLX30547-54 MFLX30547-56 MFLX30547-60
Masterflex® Tuf-Flex® Sanitary Tri-Clamp® Gaskets, Avantor®
Afdichtingen Pakkingen
Ensure a better process seal with outstanding chemical resistance.
  • The chemical resistance of PTFE and the flexibility of an elastomer
  • Designed for CIP/SIP conditions
The Tuf-Flex gasket grafts a PTFE envelope to an elastomer core. Unlike pure PTFE gaskets, or even other sheathed gaskets, Tuf-Flex will not creep or cold flow over time and therefore maintains the integrity of its seal in your process line. The core gives the gasket the mechanical properties of an elastomer, including memory and flexibility. Designed to meet the stringent requirementsMore Product Information