CPC® Twist-to-Connect Quick-Disconnect Fittings, Couplers and Accessories
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
31310-00BLK 31310-01BLK 31310-02BLK 31310-03BLK
MFLX31310-00BLKEA 198 EUR
MFLX31310-00BLK MFLX31310-01BLK MFLX31310-02BLK MFLX31310-03BLK
CPC® Twist-to-Connect Quick-Disconnect Fittings, Couplers and Accessories
Fittingen en connectoren
Twist-to-Connect locking mechanism protects against accidental disconnects.
  • Shrouded, leak-free seal protects against contamination
  • Provides rapid media transfer with smooth inner bore
These CPC® Twist-to-Connect quick-disconnect fittings feature an easy-to-use locking mechanism that guards against accidental disconnects and provides a reliable, secure connection. To assemble, place the insert in the coupling body and twist to lock. An audible click ensures your connection. A smooth inner bore and leak-free seal protect your process against contamination. All couplings are made from USP Class VI-compliant polysulfone with platinum-cured silicone seals.
A complete coupler requires a fitting body and an insert (order separately).