CPC® Quick-Disconnect Fittings, Sanitary Bodies and Inserts
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
31604-26 31604-27 31604-28 31604-30 31604-34 31604-27BLK 31604-26BLK 31604-28BLK
MFLX00296-OQEA 55.2 EUR
MFLX00296-OQ MFLX31604-26 MFLX31604-27 MFLX31604-28 MFLX31604-30 MFLX31604-34 MFLX31604-27BLK MFLX31604-26BLK MFLX31604-28BLK MFLX00296-OOBLK MFLX00296-OPBLK
CPC® Quick-Disconnect Fittings, Sanitary Bodies and Inserts
Fittingen en connectoren
Attach directly to your sanitary hardware assemblies!
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • USP Class IV materials
  • Fully sterilisable
These couplings allow you to integrate disposables with your existing stainless steel processing equipment. Both couplings and inserts are available with standard sanitary terminations to attach to your system with a sanitary gasket and tri-clover clamps. All models feature USP Class VI platinum-cured silicone gaskets.
These quick-disconnect couplings require a large fitting body and large insert (order separately).