BarbLock Ultra-Secure Tubing Retainer Assemblies
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MFLX30604-00EA 6.5 EUR
MFLX30604-00 MFLX30604-01 MFLX30604-02 MFLX30604-04 MFLX30604-06 MFLX30604-08
BarbLock Ultra-Secure Tubing Retainer Assemblies
Fittingen en connectoren
Get leak-free connection to barbed fittings.
  • Easy to install, two-piece locking mechanism ensures the tubing and hose barb won't disengage
  • Safe to use with pressures up to 140 psi or upper pressure limit of tubing
  • An excellent alternative to securing tubing and hose barbs with cable ties - no more crimps or tube distortion
BarbLock assemblies provide uniform 360-degree radial crimping for a leak-free, durable seal. The BarbLock assembly consists of three pieces: the sleeve, collet, and barbed fittings. To install the fitting, slide the sleeve andMore Product Information