Masterflex® Press-In Plug Fittings, Hose Barb, Straight, Avantor®
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MFLX31220-11EA 9.3 EUR
MFLX31220-11 MFLX41519-36 MFLX41519-44 MFLX51519-38 MFLX51519-46 MFLX41519-35 MFLX41519-43 MFLX40615-38 MFLX40615-39 MFLX40615-40 MFLX40615-41 MFLX40615-42 MFLX40615-44 MFLX40615-45 MFLX40615-43
Masterflex® Press-In Plug Fittings, Hose Barb, Straight, Avantor®
Fittingen en connectoren
Protect against contaminants in tubing lines.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include