Masterflex® Reducer Fittings, Male NPT Threaded, Straight, Avantor®
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
31701-88 31701-89
MFLX40623-35EA 28.9 EUR
MFLX40623-35 MFLX40623-43 MFLX40623-51 MFLX40623-55 MFLX40627-35 MFLX40627-43 MFLX30623-34 MFLX30623-36 MFLX30623-40 MFLX30623-50 MFLX30623-52 MFLX30623-54 MFLX30627-54 MFLX31701-88 MFLX31701-89 MFLX30623-32 MFLX30623-38 MFLX30623-42 MFLX30623-46 MFLX30623-48 MFLX40623-39 MFLX40623-47 MFLX40627-55 MFLX40615-31 MFLX40615-32 MFLX40615-33 MFLX40615-34 MFLX40615-35
Masterflex® Reducer Fittings, Male NPT Threaded, Straight, Avantor®
Fittingen en connectoren
Quick and easy assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Threaded fittings provide a more permanent fluid path. Use these fittings to reduce or enlarge connection sizes. Choose the size and material to meet your application needs.