Saint Gobain® Plastic Luer-to-Sanitary Adapters
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
31200-50 40619-96
MFLX31200-50EA 52 EUR
MFLX31200-50 MFLX40619-96
Saint Gobain® Plastic Luer-to-Sanitary Adapters
Fittingen en connectoren
Attach with just a twist!
  • Choose between polypropylene (PP) and PVDF material options
Standard connection type for syringes, plus ideal for many applications with small tubing. These Female luer lock x 3/4" mini sanitary adapters are available in either polypropylene or PVDF.