Stainless Steel Male Threaded Elbow to Push-to-Connect Adapters
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
31370-06 31370-07 31370-08
MFLX31370-06EA 74.2 EUR
MFLX31370-06 MFLX31370-07 MFLX31370-08
Stainless Steel Male Threaded Elbow to Push-to-Connect Adapters
Fittingen en connectoren
The ease of push-to-connect and the durability of 316 stainless steel.
  • Constructed of durable stainless steel
The durable stainless steel construction makes these fittings ideal for harsh environments that demand chemically resistant fluid lines. Whether required for a process area with frequent detergent washdowns or exposure to highly corrosive chemicals, these fittings ensure uninterrupted air or fluid lines. Matching material of construction to your stainless piping will reduce the effects of thermal expansion differentials and minimize line leakage.