Hedland Flow Meters for Water and Other Liquids
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
32509-75 32509-86 32509-87 32509-56
MFLX32509-75EA 498 EUR
MFLX32509-75 MFLX32509-86 MFLX32509-87 MFLX32509-56
Hedland Flow Meters for Water and Other Liquids
Debietmeters
Easy to read scale.
  • Direct reading
  • Install in any position
  • 360º Rotatable Guard/Scale
  • Easier ot Read Linear Scale
  • No Flow Straighteners or Special Piping Required
  • Insensitive to Shock and Vibration
  • Good Viscosity Stability
  • Temperature up to 240ºF
  • Accuracy ±2% Full Scale
  • Repeatability ±1%
  • Special Scales Available
Ideal for applications that are sensitive to shock and vibration.