+GF+ Signet Insertion Magmeter Flow Sensors
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
32486-49 32486-51
MFLX32486-49EA 2170 EUR
MFLX32486-49 MFLX32486-51
+GF+ Signet Insertion Magmeter Flow Sensors
Debietmeters
Accurate measurement even in dirty liquids.
  • Bi-directional flow
  • No moving parts
  • No pressure drop
These Signet magmeters are an insertion-style magnetic flow sensor that features no moving parts. The unique sensor design provides long-term reliability with minimal maintenance. Typical applications include flow monitoring and control in chemical processing, water and wastewater management, pool and spa monitoring, irrigation control, and water distribution. Use together with the Signet installation fittings on page 747 for automatic sensor alignment andMore Product Information