Masterflex® Plug Fittings, Male NPT Threaded, Straight, Avantor®
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
31411-05 31411-06 31411-08 31701-90 31701-91 31701-92
MFLX30401-38EA 28.5 EUR
MFLX30401-38 MFLX30401-39 MFLX30401-41 MFLX30401-42 MFLX30401-43 MFLX30401-44 MFLX30401-45 MFLX30401-47 MFLX30401-51 MFLX30401-52 MFLX31411-05 MFLX31411-06 MFLX31411-08 MFLX31701-90 MFLX31701-91 MFLX31701-92 MFLX30401-36 MFLX30401-40 MFLX30401-46 MFLX30401-48 MFLX30401-53
Masterflex® Plug Fittings, Male NPT Threaded, Straight, Avantor®
Fittingen en connectoren
Deigned for permanent connection.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Very good chemical resistance. Rated to 200 psi at 77 °F ( 25 °C). Usable from -65 to 190 °F (-53 to 87 °C).