Stainless Steel Compression to NPT(M) Adapters
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
31406-34 31406-35 31406-36 31406-37 31406-64 31406-65 31406-66
MFLX31406-34EA 27 EUR
MFLX31406-34 MFLX31406-35 MFLX31406-36 MFLX31406-37 MFLX31406-64 MFLX31406-65 MFLX31406-66
Stainless Steel Compression to NPT(M) Adapters
Fittingen en connectoren
Excellent choice to connect rigid tubing.
  • Great chemical resistance
  • Easy to assemble the fitting to the tubing
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through the included sealed ferrule and using the nut to compress down to form a secure seal.
Very good chemical resistance. Pressure limitation determined by the tubing, not the fitting. Usable from -40 to 200ºF (-40 to 93ºC). Sterilize by either ethylene oxide, autoclave, or gamma irradiation.