CPC® Quick-Disconnect Fittings, Hose Barb Bodies
Leverancier: CPC (Colder Products Company)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MFLX06364-65BLKEA 395 EUR
MFLX06364-65BLK MFLX06360-10 MFLX06360-55 MFLX06361-51BLK MFLX06364-19 MFLX06360-15 MFLX06360-12 MFLX06360-57 MFLX06364-15 MFLX06361-03BLK MFLX31307-04 MFLX06364-10 MFLX06364-55 MFLX06360-10BLK MFLX06360-65BLK MFLX06364-55BLK MFLX06361-12BLK MFLX06360-19 MFLX06361-12 MFLX06361-11 MFLX06360-55BLK MFLX06361-52BLK MFLX06361-53 MFLX06364-70BLK MFLX06361-13 MFLX06361-04BLK MFLX06364-10BLK MFLX06361-52 MFLX06361-51 MFLX06360-20BLK MFLX06364-25BLK MFLX06361-11BLK MFLX06361-46BLK MFLX06364-09 MFLX06360-09 MFLX06361-53BLK MFLX06364-15BLK MFLX06361-05 MFLX06361-04 MFLX06364-19BLK MFLX06361-48 MFLX06361-03 MFLX06361-05BLK MFLX06361-46 MFLX06364-70 MFLX06360-12BLK MFLX06360-70 MFLX06360-67BLK MFLX06361-09 MFLX06364-09BLK MFLX06364-60BLK MFLX06360-25BLK MFLX06364-20BLK MFLX06360-65 MFLX06360-57BLK MFLX06360-22 MFLX06360-20 MFLX06360-25 MFLX06360-67 MFLX06364-25 MFLX06364-20 MFLX06364-65 MFLX06364-60 MFLX06360-15BLK MFLX06360-60 MFLX06360-19BLK MFLX06360-70BLK MFLX06360-60BLK MFLX06361-48BLK MFLX06360-09BLK MFLX06360-22BLK
CPC® Quick-Disconnect Fittings, Hose Barb Bodies
Fittingen en connectoren
No more struggling - just press thumb latch.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Fittings connect in a snap
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. The audible click when the coupling engages confirms a solid connection. To disconnect fitting halves, simply press the button on the coupling body. Standard flow quick-disconnect couplings require a body and an insert of the same flow size.
A complete coupler requires a fitting body and an insert (order separately).