High-Pressure Diaphragm Pumps and Integrated Coolant Systems
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
75421-11 75421-02 75421-12
MFLX75421-11EA 3710 EUR
MFLX75421-11 MFLX75421-02 MFLX75421-12
High-Pressure Diaphragm Pumps and Integrated Coolant Systems
Pompen Diaphragm Pumps
Worry-free pumping of common industrial fluids.
  • High-performance diaphragm pumps handle a wide range of industrial fluids
  • Liquid end and motor/drive withstands high discharge pressures up to 1000 psi
  • Brass or 316 stainless steel (SS) pump head
These high-performance diaphragm pumps can handle a wide range of industrial fluids—from liquid butane to drinking water to ceramic slurries. Whatever the application, you can be assured these rugged diaphragm pumps get the job done without worrying about pump failure or costly repairs.
To ensure yourMore Product Information