Masterflex® Peristaltic Pump Footswitches, Avantor®
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MFLX07523-92EA 447 EUR
MFLX07523-92 MFLX07575-84 MFLX07595-43
Masterflex® Peristaltic Pump Footswitches, Avantor®
Pompen Peristaltische pompen
Easily control your Masterflex® drive with compatible footswitch.
  • Choose footswitch with the correct connection for your Masterflex® pump
Add convenience to repetitive dispensing with a foot-operated on/off switch. Keep hands free to efficiently move containers during routine fill applications or to keep containers stable during the filling process.