Masterflex® Remote Control Cable Connectors, Avantor®
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MFLX07523-94EA 159 EUR
MFLX07523-94 MFLX07595-45 MFLX07595-52
Masterflex® Remote Control Cable Connectors, Avantor®
Kabels
Create your own cable for optimization of your Masterflex® drive.
  • Choose the correct connection for your Masterflex® pump
Combine these connectors with 20-22 gauge shielded wire to create your own interface cable. Maximum cable length is 1000 ft (300 m).