Masterflex® Smart Gasket® Sanitary Gasket Accessories, Avantor®
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MFLX30545-98EA 103 EUR
MFLX30545-98 MFLX30545-99
Masterflex® Smart Gasket® Sanitary Gasket Accessories, Avantor®
Afdichtingen Pakkingen
Optimize performance of your Masterflex® Smart Gasket® Sanitary Gasket system.
  • Choose from a wide range of replacement parts and accessories
Smart Gaskets allow you to measure temperature, take samples, or dose chemicals in your process line. Selected accessories include thermocouple probes and wire, port plugs for the Smart Gasket, and a sampling tube.