TriClamp Sanitary Strainer Assemblies, 316 Stainless Steel
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
29590-02 29590-14
MFLX29590-02EA 1390 EUR
MFLX29590-02 MFLX29590-14
TriClamp Sanitary Strainer Assemblies, 316 Stainless Steel
Filters Strainers
Protect products and expensive process equipment from coarse or fine particulates.
  • 316 SS fluid contact surfaces are polished to a smooth, blemish free finish
  • Standard clamp sizes allow easy installation or retrofit
Sanitary strainers are ideal for particulate removal or pre-filtration in the processing of foods, beverages, pharmaceuticals, cosmetics, and other critical compounds. Housings are available in in-line and side-inlet configurations.
Leveringsinformatie: Strainer assemblies include housing, perforated core, internal cap and spring,More Product Information