GF Signet® Manual Diaphragm Valves, PVC
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
07390-50 07390-54 07390-56
MFLX07390-50EA 343 EUR
MFLX07390-50 MFLX07390-54 MFLX07390-56
GF Signet® Manual Diaphragm Valves, PVC
Ventielen Diaphragm Valves
Optimized valve body provides double the CV values.
  • Two-color positition indicator on handwheel
  • Elimination of bolts removes corrosive weak points
  • Mount vertically or horizontally
  • PVC body and EPDM diaphragm
These GF Signet® manual diaphragm valves are constructed with a PVC body and EPDM diaphragm. Valves offer a maximum temperature of 150 °F (65 °C) and feature true union socket connections.