Masterflex® Sani-Lock 316L Stainless Steel Quick-Connect Clamps, Avantor®
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
31607-70_U 31607-84_U
MFLA31607-70_UEA 0 EUR
MFLA31607-70_U MFLA31607-84_U MFLX31607-70 MFLX31607-76 MFLX31607-84
Masterflex® Sani-Lock 316L Stainless Steel Quick-Connect Clamps, Avantor®
Fittingen en connectoren
No hinge clamp required – rapid one-hand connection.
  • Snap-in-place locking arms secure fitting, while safety pins prevent accidental release
  • Fully compatible with sanitary Tri-Clamp® fittings
  • 150 psi (10.3 bar) working line pressure
Converts any connection into a sanitary 'quick-connect' connection. Sani-Lock provides the convenience and efficiency of a quick-connect while maintaining the integrity of a sanitary connection. Use with Tuf-Flex® gaskets for guaranteed leak-proof alignment.(sold separately below) Automatic alignment of the Tuf-Flex gasketMore Product Information