Hand-Operated Siphon Drum Pump Accessories
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
07079-10 07079-15 07079-17
MFLX07079-10EA 30.1 EUR
MFLX07079-10 MFLX07079-15 MFLX07079-17
Hand-Operated Siphon Drum Pump Accessories
Pompen Vatpompen
Compatible with Siphon and Piston Hand-Operated Pumps.
  • Choose from a wide range of replacement parts and accessories
Model 07079-10 steel screw in adapter features a 1-3/4" NPT(F) connection and fits 5 gallon pails.
Model 07079-15 flex spout adapter is constructed of durable polyethylene and fits 5 gallon containers with 5-5/8" (6.7 cm) openings.
Model 07079-17 bung converter kit converts a Mauser course bung thread or standard course buttress to a 2" NPT connection.