Tygon® E-1000 Ultra-Soft Flexible Transfer Tubing
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
65102-00 65102-02 65102-04 65102-06 65102-08 65102-16 65102-10 65102-14
MFLX65102-00EA 169 EUR
MFLX65102-00 MFLX65102-02 MFLX65102-04 MFLX65102-06 MFLX65102-08 MFLX65102-16 MFLX65102-10 MFLX65102-14
Tygon® E-1000 Ultra-Soft Flexible Transfer Tubing
Slangen Transfer Tubing
High degree of flexibility, performs well at low temperatures.
  • Global market leader in range of formulations for most extensive chemical compatibility
Formulation: Tygon® E-1000
Applications: General laboratory, vent and drain lines. Ideal for lab installations requiring multiple directional changes, abrupt curves, or twisting.
Characteristics: High degree of flexibility, performs well at low temperatures, broad chemical resistance, resists kinking and collapse. Transparent.
Certification: FDA compliant. Made according to GMP.
Temperature range: –67 to 125 °F (–55 to 52 °C)
Sterilize:Autoclave or ethylene oxide.